Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Volatiles MegaMix™ with Gases, Restek
SDP

Catalog No. 08776254 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-776-254 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-776-254 Supplier Restek Corporation Supplier No. 30603
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Volatiles MegaMix with Gases
Density 1.1 (air = 1)
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Each component at 200μg/mL in P&T methanol (60 components)
benzene
bromobenzene
bromochloromethane
bromodichloromethane
bromoform
bromomethane (methyl bromide)
n-butylbenzene
sec-butylbenzene
tert-butylbenzene
carbon tetrachloride
chlorobenzene
chloroethane (ethyl chloride)
chloroform
chloromethane (methyl chloride)
2-chlorotoluene
4-chlorotoluene
dibromochloromethane
1,2-dibromo-3-chloropropane (DBCP)
1,2-dibromoethane (EDB)
dibromomethane
1,2-dichlorobenzene
1,3-dichlorobenzene
1,4-dichlorobenzene
dichlorodifluoromethane (CFC-12)
1,1-dichloroethane
1,2-dichloroethane
1,1-dichloroethene
cis-1,2-dichloroethene
trans-1,2-dichloroethene
1,2-dichloropropane
1,3-dichloropropane
2,2-dichloropropane
1,1-dichloropropene
cis-1,3-dichloropropene
trans-1,3-dichloropropene
ethylbenzene
hexachloro-1,3-butadiene (hexachlorobutadiene)
isopropylbenzene (cumene)
4-isopropyltoluene (p-cymene)
methylene chloride (dichloromethane)
naphthalene
n-propylbenzene
styrene
1,1,1,2-tetrachloroethane
1,1,2,2-tetrachloroethane
tetrachloroethene
toluene
1,2,3-trichlorobenzene
1,2,4-trichlorobenzene
1,1,1-trichloroethane
1,1,2-trichloroethane
trichloroethene
trichlorofluoromethane (CFC-11)
1,2,3-trichloropropane
1,2,4-trimethylbenzene
1,3,5-trimethylbenzene
vinyl chloride
m-xylene
o-xylene
p-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.