Learn More
Vee Gee Scientific, Inc. Volumetric Flask, Class A, Snap Cap, 1000 mL
Supplier:  Vee Gee Scientific, Inc. 2303A1000
Encompass_Preferred
The SIBATA Class A Volumetric Flask features machine-blown body and heavy-beaded, heavy-tubing neck that is tooled for a snap-cap fit.• Designed """"To Contain"""" with a temperature of calibration at 20°C• Constructed of borosilicate glass with a low coefficient of expansion• Graduation line is sharply defined and of uniform width• Flask has a large marking square and includes a snap-cap closure• Meets ASTM E-288 Class A Specifications & USP Standards for Volumetric Flasks
Catalog No. 50-225-0370
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.