Learn More
Eisco™ Volumetric Flask with Stopper
Catalog No. 12183048 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
200 mL
500 mL
1000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-183-048 25 mL
12-183-045 50 mL
12-183-044 100 mL
12-183-043 200 mL
12-183-040 500 mL
12-183-039 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 12-183-048 Supplier Eisco™ Supplier No. CH1952DST
Volumetric Flask, Class A with Polypropylene Stopper and a blue graduation. Made of Borosilicate Glass.
  • Made of high quality, heavy duty, borosilicate 3.3 glass
  • Class A
  • Single, white graduation mark
  • Includes interchangeable polypropylene stopper
Specifications
Capacity (Metric) 25 mL
Material Borosilicate Glass (3.3)
Product Type Volumetric Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Beaded
Sterility Non-sterile
Class Class A
Bottom Shape Flat
Surface Treatment Uncoated