Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Hippel Lindau Antibody (1066402) [DyLight 350], Novus Biologicals™
SDP

Catalog No. FAB11435D
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB11435D 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB11435D Supplier R&D Systems Supplier No. FAB11435D

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Von Hippel Lindau Monoclonal antibody specifically detects Von Hippel Lindau in Human samples. It is validated for Western Blot

Specifications

Antigen Von Hippel Lindau
Applications Western Blot
Classification Monoclonal
Clone 1066402
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias elongin binding protein, HRCA1, Protein G7, pVHL, RCA1, VHL1, von Hippel-Lindau disease tumor suppressor, von Hippel-Lindau syndrome, von Hippel-Lindau tumor suppressor
Host Species Mouse
Immunogen E. coli -derived recombinant human VHL, Asn150-Asp213, Accession # P40337
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, HIF Target Genes, Hypoxia, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7428
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.