Learn More
StatLab VON KOSSA STAIN KIT
SDP
Supplier:  StatLab KTVKO
Encompass_Preferred
Kit, Von Kossa Stain; Stains for Calcium Deposits; Contains 5% Silver Nitrate 100mL, 5% Sodium Thiosulfate 100mL and Nuclear Fast Red Stain 100mL; Kit is sealed with a tamper-evident label
Catalog No. NC9239431
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.