Learn More
Biomatik Corporation Von Willebrand Factor (Vwf) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30063
Encompass_Preferred
Von Willebrand Factor (Vwf) Monoclonal Antibody is a Rat (Rattus norvegicus) Monoclonal Antibody to Von Willebrand Factor (vWF). The Von Willebrand Factor (Vwf) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Rat (Rattus norvegicus). Immunogen: Lys49~Asn230. Applications: WB. Target Synonyms: Von Willebrand Factor (vWF). Accession Number: Q62935. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5238
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.