Learn More
Selleck Chemical LLC Vorasidenib AG-881-2mg
SDP
Supplier:  Selleck Chemical LLC S86112MG
Encompass_Preferred
Vorasidenib (AG-881) is an orally available inhibitor of mutated forms of both isocitrate dehydrogenase 1 and 2 (IDH1 and IDH2).
Catalog No. 50-238-9794
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.