Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Voriconazole, Anti-Fungal Agent, Solid, >99% Purity, 10 mg

Catalog No. 502706937
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6937 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6937 Supplier Abcam Supplier No. AB14106710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent and broad-spectrum anti-fungal agent with >99% purity.

  • Purity: >99%
  • Molecular Weight: 349.31 Da
  • Mechanism: Inhibits CYP450-dependent 14-α sterol demethylase to interfere with ergosterol biosynthesis and inhibit fungal growth
  • Form: Solid
  • Active: In vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule 14-α Sterol Demethylase
Shipping Condition Ambient
Alternate Part Number UK-109, 496
Buffer 100% Voriconazole
Content And Storage Store at 4°C.
Form Solid
Formulation 100% Voriconazole
Quantity 10 mg
For Use With (Application) Inhibition of fungal growth
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.