Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Voriconazole

Catalog No. 376010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-601-0 10 mg
37-605-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-601-0 Supplier Tocris Bioscience™ Supplier No. 3760/10
This item is not returnable. View return policy

Triazole antifungal agent

Triazole antifungal agent. Displays potent activity against B87964, Cryptococcus and Aspergillus species.

Chemical Identifiers

CAS 137234-62-9
Molecular Formula C16H14F3N5O

Specifications

Quantity 10 mg
Formula Weight 349.31
Purity >99%
Molecular Weight (g/mol) 349.317
Chemical Name or Material Voriconazole
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.