Learn More
Scientific Industries VORTEX-GENIE 1 TOUCH 230V NP
Supplier:  Scientific Industries SI0146
Encompass_Preferred
Violent high-speed TOUCH mixer attacks problem applications. Specially designed feet and heavy casting prevents walking. 6.0mm orbit ideal for Touch Mixing. Does not include a plug.
Catalog No. 50-728-014
Only null left
Add to Cart
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.