Learn More
Corning™ LSE™ Digital Vortexer Platform
Catalog No. 07202206 Shop All Corning Products
Change view
Click to view available options
Product Type:
Platform
Platform with non-stick mat
Spring Loaded Platform
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
07-202-206 Platform
07-202-207 Platform with non-stick mat
07-202-208 Spring Loaded Platform
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-202-206 Supplier Corning™ Supplier No. 48010
Add to Cart
Add to Cart
Corning™ Vortexer Platform for use with Corning™ LSE Multiposition Digital Vortexer.
The Corning Digital Vortexer is a multipurpose shaker dedicated to a variety of shaking and mixing applications. Three easily removable and interchangeable platforms accommodate many different sample containers such as flasks, bottles, tube racks, plates, boxes, and dishes. The clamp platform has spring loaded retaining bars that adjust to fit vessels of different shapes and sizes. The microplate platform is designed to securely hold up to four microplates. Operation of the vortexer can be timed or continuous, while the speed is variable. The broad speed range allows the vortexer to be used for gentle mixing as well as vigorous agitation. A powerful motor, and a chemically resistant and easy to clean surface makes the equipment a perfect choice for years of trouble-free use.
Features and Benefits
  • Three versatile platform options (available separately)
  • Compact design with easy-access control panel
  • Non-slip rubber mat to keep containers in place
  • Safe for cold room or incubator use
  • Five-year warranty
TRUSTED_SUSTAINABILITY
Specifications
Holds 4 Microplates
Product Type Platform
Description Designed to securely hold up to four microplates
For Use With (Application) General Purpose Mixing
For Use With (Equipment) LSE Multiposition Digital Vortexer
No. of Positions 4
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov