Learn More
Apexbio Technology LLC Vortioxetine (Lu AA21004) HBr 960203-27-4 100mg
Supplier: Apexbio Technology LLC A2547100
Vortioxetine (Lu AA21004) HBr (CAS 960203-27-4) is a multimodal antidepressant that modulates the serotonergic system It acts as an antagonist at the human 5-HT3A (Ki 3 7 nM) and 5-HT7 receptors (Ki 19 nM) a partial agonist at the 5-HT1B receptor (Ki 33 nM EC50 460 nM) an agonist at 5-HT1A (Ki 15 nM) and inhibits the serotonin transporter (hSERT Ki 1 6 nM) In cellular assays it demonstrates functional antagonism at 5-HT7 and partial agonism at 5-HT1B In vivo vortioxetine modulates serotonergic activity in rodents with ED50 values of 0 4 mg/kg for rSERT and 0 11 mg/kg for 5-HT3 It is utilized in neuropharmacological studies to investigate serotonergic signaling and depression-related mechanisms
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.