Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPARP Monoclonal Antibody (P193-10)
GREENER_CHOICE

Catalog No. 01672051
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-051 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-051 Supplier Invitrogen™ Supplier No. MA135745
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p193-10 reacts with an internal epitope (amino acids 506-510, VALGK) of the minor vault protein (p193 or VPARP), which is overexpressed in various human non-P-glycoprotein MDR tumor cell lines, accordingly to an increase in the number of vault particles. p193-10 was raised against an E. coli lysate transformed with the pET28a(+) expression vector containing amino acids 408-611 of the p193 cDNA.

This gene encodes poly(ADP-ribosyl)transferase-like 1 protein, which is capable of catalyzing a poly(ADP-ribosyl)ation reaction. This protein has a catalytic domain which is homologous to that of poly (ADP-ribosyl) transferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly (ADP-ribosyl) transferase. Since this protein is not capable of binding DNA directly, its transferase activity may be activated by other factors such as protein-protein interaction mediated by the extensive carboxyl terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPARP
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone P193-10
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide
Gene PARP4
Gene Accession No. Q9UKK3
Gene Alias 193 kDa vault protein; ADP-ribosyltransferase (NAD+; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 1; ADP-ribosyltransferase diphtheria toxin-like 4; ADPRTL1; ARTD4; C030027K23Rik; E230037B21Rik; Gm743; H5 proline-rich; I-alpha-I-related; KIAA0177; p193; Parp4; PARP-4; PARPL; PARP-related; PARP-related/IalphaI-related H5/proline-rich; PH5P; poly (ADP-ribose) polymerase family, member 4; poly (ADP-ribose) polymerase)-like 1; poly [ADP-ribose] polymerase 4; poly(ADP-ribose) polymerase family member 4; poly(ADP-ribose) synthetase; poly(ADP-ribosyl)transferase-like 1; Protein mono-ADP-ribosyltransferase PARP4; Vault poly(ADP-ribose) polymerase; vault protein, 193-kDa; VAULT3; von Willebrand factor A domain containing 5C; VPARP; VWA5C
Gene Symbols PARP4
Host Species Mouse
Immunogen VPARP aa 408-611.
Purification Method purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.