Learn More
Medchemexpress LLC (S)-N-(1-(3,4-difluorophenyl)ethyl)-1H-indazole-3-carboxamide | 877969-69-2 | 99.0% | 301.29 g/mol | C16H13F2N3O | 25MG
SDP
Supplier:  Medchemexpress LLC HY12607625MG
Encompass_Preferred
VPC-80051 racemate is the racemic form of VPC-80051, a small-molecule inhibitor of the hnRNP A1 splicing factor. It directly interacts with the hnRNP A1 RNA-binding domain and has been reported to reduce AR-V7 messenger RNA levels in 22Rv1 castration-resistant prostate cancer cells. Intended for research use only and supplied as a white to off-white solid.
  • Inhibits hnRNP A1 splicing activity
  • Reduces AR-V7 mRNA in 22Rv1 cells
  • Supplied as a white to off-white powder for laboratory use
  • High purity suitable for research applications
  • Stable when stored at recommended low temperatures
Catalog No. 50-002-38302
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.