Learn More
Meso Scale Discovery V-PLEX Viral Panel 2 NHP Kit
SDP
Supplier:  Meso Scale Discovery K15349D2
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 2 (NHP) Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum and plasma samples This assay utilizes MSD s electrochemiluminescence detection technology and provides high sensitivity up to a 7-log dynamic range and reliable results from small sample volumes It includes a 96-well plate pre-coated with antigens detection antibodies buffers calibrators and controls needed to run the 4-plex assay for up to 80 tests This kit enables efficient viral antibody profiling from non-human primate studies
Catalog No. 50-000-78271
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.