Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 28 IgA
SDP
Supplier:  Meso Scale Discovery K15616U4
Encompass
The Meso Scale Discovery COVID-19 Spike Protein Neutralization Assay Kit is a sensitive immunoassay for measuring neutralizing antibodies against SARS-CoV-2 in serum and plasma It uses recombinant spike protein on plates to capture antibodies with a detection antibody emitting an electrochemiluminescent signal measured by Meso Scale Discovery instruments The assay detects antibodies at the pg/mL level providing quantitative data on antibody levels The kit includes all necessary reagents and processes up to 80 samples per plate
Catalog No. 50-000-80962
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.