Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPS16 Isoform 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537963
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537963 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537963 Supplier Invitrogen™ Supplier No. PA537963
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS16 Isoform 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene VPS16
Gene Accession No. Q920Q4, Q9H269
Gene Alias 1810074M16Rik; hVPS16; mVPS16; RP11-12M19.2; vacuolar protein sorting 16 (yeast); vacuolar protein sorting 16 homolog; vacuolar protein sorting 16 homolog (S. cerevisiae); vacuolar protein sorting protein 16; vacuolar protein sorting-associated protein 16 homolog; VPS16; VPS16 CORVET/HOPS core subunit; VPS16, CORVET/HOPS core subunit
Gene Symbols VPS16
Host Species Goat
Immunogen Peptide with sequence C-KESQKADEYLREIQ, from the internal region of VPS16 (aa 358-371).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296159, 64601, 80743
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.