Learn More
Apexbio Technology LLC VPS34-IN1 1383716-33-3 100mg
SDP
Supplier:  Apexbio Technology LLC B6179100
Encompass_Preferred
VPS34-IN1 (CAS 1383716-33-3) is a highly selective small molecule inhibitor of the class III phosphoinositide 3-kinase Vps34 exhibiting an in vitro IC50 of 25 nM Vps34 is responsible for phosphorylation of phosphatidylinositol to generate PtdIns(3)P which coordinates membrane trafficking through recruitment of PX and FYVE domain-containing proteins VPS34-IN1 potently inhibits Vps34 without significantly affecting more than 340 protein kinases or 25 lipid kinases including all class I and II PI3K isoforms Cellular studies show VPS34-IN1 induces rapid dose-dependent dispersion of PtdIns(3)P-binding probes and diminishes SGK3 phosphorylation aiding in the elucidation of Vps34 signaling and PI3K pathway interactions in cellular models
Catalog No. 50-000-92134
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.