Learn More
Invitrogen™ VPS51 Polyclonal Antibody

Description
Recommended positive controls: A431, HeLa, HepG2. Predicted reactivity: Mouse (99%), Zebrafish (88%), Xenopus laevis (83%), Chicken (80%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | VPS51 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.26 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | VPS51 |
| Gene Accession No. | Q9UID3 |
| Gene Alias | 1110014N23Rik; 3110057M17Rik; AI837850; ANG2; ANG3; Another new gene 2 protein; C11orf2; C11orf3; Ffr; PP5382; Protein fat-free homolog; vacuolar protein sorting 51 homolog; vacuolar protein sorting 51 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 51 homolog; VPS51; VPS51 GARP complex subunit; VPS51, GARP complex subunit |
| Gene Symbols | VPS51 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.