Learn More
Invitrogen™ VPS52 Polyclonal Antibody

Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | VPS52 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | VPS52 |
| Gene Accession No. | Q8N1B4 |
| Gene Alias | Are1; D130068D18; D430041K17Rik; dJ1033B10.5; DKFZp547I194; fb94c11; LOW QUALITY PROTEIN: vacuolar protein sorting-associated protein 52 homolog; RP5-1033B10; SAC2; SAC2 suppressor of actin mutations 2-like protein; SACM2L; tclw5; vacuolar protein sorting 52 (yeast); vacuolar protein sorting 52 homolog; vacuolar protein sorting 52 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 52 homolog; vacuolar protein sorting-associated protein 52-like protein; Vacuolar protein sorting-associated protein 52-like protein-like protein; Vps52; VPS52 GARP complex subunit; VPS52 subunit of GARP complex; VPS52, GARP complex subunit; wu:fb94c11 |
| Gene Symbols | VPS52 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.