Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolyCo™ VR™ Protective Gowns, Elastic Cuffs

Catalog No. 19125519
Change view
Click to view available options
Color:
Blue
White
Yellow
Size:
Large
Regular
X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-125-519 Yellow Regular
19-125-518 Blue Regular
19-125-583 White X-Large
19-125-522 Blue X-Large
19-125-523 Yellow X-Large
19-125-521 Yellow Large
19-125-520 Blue Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-125-519 Supplier PolyCo™ Supplier No. 42455
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gowns provide greater coverage for applications where the threat of biological or chemical contamination exist.

  • Glove-to-sleeve protection
  • Full-frontal coverage suitable for workplaces with biological or chemical hazards
  • Slips on over workers' heads and ties securely in the back for a quick and easy fit
  • Secures sleeves in place with elastic wrists
  • Durable 6 mil thickness

Specifications

Size Regular
Gender Unisex
Length (English) 45 in.
Length (Metric) 114.3 cm
For Use With (Application) Food Processing, Industrial, Manufacturing
Product Type General Purpose Gown
No. of Pockets None
Packaging Type Bulk Packaged
Material Polyolefin Plastomer Resins
Color Yellow
Closure Type Back Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Model 42455
Seam Type Welded
Sleeve Style Long Sleeve
Thickness 6 mil
Certifications/Compliance OSHA 29, CFR Part 1910.1030
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.