Learn More
PolyCo™ VR™ Protective Gowns with Thumb Loops
Catalog No. 19125510
This item is not returnable. View return policy
Catalog No. 19-125-510 Supplier PolyCo™ Supplier No. 42402
Add to Cart
Add to Cart
This item is not returnable. View return policy
Gowns provide greater coverage for applications where the threat of biological or chemical contamination exist.
  • Glove-to-sleeve protection
  • Full-frontal coverage suitable for workplaces with biological or chemical hazards
  • Slips on over workers' heads and ties securely in the back for a quick and easy fit
  • Secures sleeves in place with thumb loop
  • Durable 4 mil thickness
Specifications
Length (English) 45 in.
Length (Metric) 114.3 cm
For Use With (Application) Food Processing, Critical and Controlled Environments, Healthcare, Manufacturing
Product Type Controlled Environments Gown
Closure Type Ties
Sterility Non-sterile
Seam Type Welded
Sleeve Style Full Sleeve with Thumb Loop
Certifications/Compliance OSHA 29, CFR Part 1910.1030