Learn More
PolyCo™ VR™ Protective Gowns with Thumb Loops
Description
- Glove-to-sleeve protection
- Full-frontal coverage suitable for workplaces with biological or chemical hazards
- Slips on over workers' heads and ties securely in the back for a quick and easy fit
- Secures sleeves in place with thumb loop
- Durable 4 mil thickness
Specifications
Specifications
| Length (English) | 50 in. |
| Length (Metric) | 127 cm |
| For Use With (Application) | Food Processing, Critical and Controlled Environments, Healthcare, Manufacturing |
| Product Type | Controlled Environments Gown |
| Closure Type | Ties |
| Sterility | Non-sterile |
| Seam Type | Welded |
| Sleeve Style | Full Sleeve with Thumb Loop |
| Certifications/Compliance | OSHA 29, CFR Part 1910.1030 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.