Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VRK2 Mouse anti-Human, Clone: OTI2F5, liquid, TrueMAB™
SDP

Catalog No. 501684011
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4011 Supplier OriGene Supplier No. TA811397
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a member of the vaccinia-related kinase family of serine/threonine protein kinases. This gene is widely expressed in human tissues and has increased expression in actively dividing cells, such as those in testis, leukocytes, fetal liver, and carcinomas. Its protein localizes to the endoplasmic reticulum and has been shown to phosphorylate casein and undergo autophosphorylation. Multiple alternatively spliced transcript variants have been found for this gene.

Specifications

Antigen VRK2
Applications Western Blot
Classification Monoclonal
Clone OTI2F5
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene VRK2
Gene Accession No. Q86Y07
Gene Alias vaccinia related kinase 2
Gene Symbols VRK2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 301-486 of human VRK2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7444
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.