Learn More
ORS Nasco VS130HV AMERICAS VERISHIELD EA
Supplier:  ORS Nasco 1541035110VS
Encompass_Preferred
Honeywell part# 1035110-VS. Honeywell VeriShield 100 Series Over-the-Head Ear Muffs Style: Over-the-head High-visibility yellow Noise Reduction Rating: 30 dB ANSI S3.1974: Yes CSA Z94.2: Class AL Dielectric: No Foldable: No High Visibility: Yes ABS Model: VS130HV
Catalog No. 50-255-1618