Learn More
Description
Specifications
Specifications
| Assay Range | 31.25 pg/mL |
| Assay Sensitivity | 31.25-2000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human VSIG4. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily |
| Gene ID (Entrez) | 11326 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
