Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSIG4 Monoclonal Antibody (JAV4), Super Bright™ 600, eBioscience™, Invitrogen™

Catalog No. p-7162235
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-575-742 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 63-575-742 Supplier Invitrogen™ Supplier No. 63575742
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This JAV4 monoclonal antibody recognizes human V-Set and Immunoglobulin domain containing 4 (VSIG4), also known as Complement Receptor of the Immunoglobulin superfamily (CRIg) or Z39Ig. There are two variants of human VSIG4 that result from alternative splicing. Isoform 1 is longer and more prevalent than isoform 2, and this JAV4 antibody recognizes both. VSIG4 is a type I transmembrane glycoprotein structurally related to the B7 family of immune regulatory proteins. It contains one complete V-type Ig domain and one truncated C-type Ig domain. VSIG4 is exclusively expressed on tissue resident and tumor infiltrating macrophages. It has been shown to bind complement components C3b and iC3b. This binding inhibits the alternative complement pathway and facilitates phagocytosis of complement-opsonized pathogens. VSIG4 has also been reported to suppress T cell activation, proliferation and IL-2 production thereby playing a role in the maintece of peripheral T cell tolerance and suppression of established inflammation. Expression of VSIG4 on tumor-infiltrating macrophages suggests its role in immune evasion. Pro-inflammatory stimuli such as TNF and LPS have been reported to down-regulate the expression of VSIG4. The JAV4 antibody will recognize VSIG4 on cells that have been formaldehyde-fixed and permeabilized. This antibody does not cross-react with mouse or rat VSIG4. Applications Reported: This JAV4 antibody has been reported for use in flow cytometric analysis.

T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSIG4
Applications Flow Cytometry
Classification Monoclonal
Clone JAV4
Concentration 5 μL/Test
Conjugate Super Bright 600
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene VSIG4
Gene Accession No. Q9Y279
Gene Alias A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG
Gene Symbols VSIG4
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11326
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.