Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VSIG4 Monoclonal Antibody (NLA14), Super Bright™ 436, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 62575282
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-575-282 100 μg
62-575-280 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 62-575-282 Supplier Invitrogen™ Supplier No. 62575282
  • $478.00
    $358.65 / Each of 1

    Save $119.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: This NLA14 monoclonal antibody recognizes mouse V-Set and Immunoglobulin domain containing 4 (VSIG4), also known as Complement Receptor of the Immunoglobulin superfamily (CRIg) or Z39Ig. VSIG4 is a type I transmembrane glycoprotein structurally related to the B7 family of immune regulatory proteins. It contains one complete V-type Ig domain and one truncated C-type Ig domain. VSIG4 is exclusively expressed on tissue resident and tumor infiltrating macrophages. It has been shown to bind complement components C3b and iC3b. This binding inhibits the alternative complement pathway and facilitates phagocytosis of complement-opsonized pathogens. VSIG4 has also been reported to suppress T cell activation, proliferation and IL-2 production thereby playing a role in the maintece of peripheral T cell tolerance and suppression of established inflammation. Expression of VSIG4 on tumor-infiltrating macrophages suggests its role in immune evasion. Pro-inflammatory stimuli such as TNF and LPS have been reported to down-regulate the expression of VSIG4. Peritoneal macrophages in Balb/c mice express significantly higher levels of VSIG4 than such macrophages in C57Bl/6 or Swiss Webster mice. The NLA14 antibody will recognize VSIG4 on cells that have been formaldehyde-fixed and permeabilized. This antibody does not block the ligation of VSIG4 to its T cell ligand. The NLA14 antibody does not cross-react with rat or human VSIG4.

T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSIG4
Applications Flow Cytometry
Classification Monoclonal
Clone NLA14
Concentration 0.2 mg/mL
Conjugate Super Bright 436
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene VSIG4
Gene Alias A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG
Gene Symbols VSIG4
Host Species Rat
Immunogen Fc protein and C6 cells.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 278180
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.