Learn More
IW Tremont Co Inc Filter Binderless glass microfiber VSS 1.5um 2.5cm dia. Determine fixed & volatile solids Ignited at 550°C Method 2540E. Use with standard methods 2540C & 2540D and EPA Method 160.2 for water quality in suspended solids content 100/pk
SDP
Supplier:  IW Tremont Co Inc LEVSS2500
Encompass_Preferred
Grade VSS is a binder free material manufactured using a proprietary glass chemistry which permits usage in high heat applications beyond typical borosilicate glass blends. Ideally suited for determination of "Fixed & Volatile Solids Ignited at 550°C Method 2540E. Low fiber shedding for quality assurance of test results and low percentage of weight loss when used in gravimetric tests. Particle retention 1.50um and high loading capacity is an attribute of the high surface area and complex pore structure. Material is compliant with the requirements of standard methods 2540C & 2540D and EPA Method 160.2 for establishing water quality in suspended solids content. Also used in air pollution monitoring, high temperature flue gas and filtration of high temperature solvents. “The residue of TSS, TDS after heating to dryness for a defined amount of time and at a defined temperature is defined as Fixed solids. Volatile solids are those solids lost on ignition (heating to 550 degrees C.)"
Catalog No. 50-223-7193