Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV-Glycoprotein Mouse anti-Virus, Clone: 8G5F11, MilliporeSigma™

Catalog No. p-7222138 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF233725 25 μg
MABF2337100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF233725 Supplier MilliporeSigma Supplier No. MABF233725UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VSV-Glycoprotein Monoclonal antibody specifically detects VSV-Glycoprotein in Virus samples. It is validated for Flow Cytometry, Inhibition Assay, Neutralization, Radioimmune Assays, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSV-Glycoprotein
Applications Flow Cytometry, Inhibition Assays, Neutralization, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 8G5F11
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in PBS without azide.
Host Species Mouse
Immunogen Vesicular stomatitis virus, Indiana serotype
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.