Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VU 0240551, Tocris Bioscience™

Catalog No. 388810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-881-0 10 mg
38-885-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-881-0 Supplier R&D Systems Supplier No. 3888/10
This item is not returnable. View return policy

KCC2 inhibitor

Inhibitor of the neuronal K-Cl cotransporter, KCC2 (IC50 = 560 nM for K+ uptake assay in KCC2-overexpressing cells). Exhibits selectivity over the Na-K-2Cl cotransporter, NKCCl. Also inhibits hERG and L-type Ca2+ channels.

Chemical Identifiers

CAS 893990-34-6
Molecular Formula C16H14N4OS2
Molecular Weight (g/mol) 342.435
InChI Key WJRWSLORVIHRNX-UHFFFAOYSA-N
Synonym n-4-methyl-2-thiazolyl-2-6-phenyl-3-pyridazinyl thio-acetamide, n-4-methyl-2-thiazolyl-2-6-phenyl-3-pyridazinyl thio acetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl thio acetamide, n-4-methylthiazol-2-yl-2-6-phenylpyridazin-3-ylthio acetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl sulfanylacetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl sulfanyl acetamide, d0m2pp, cid, vu hplc, n-4-methylthiazol-2-yl-2-6-phenylpyridazin-3-yl thio acetamide
PubChem CID 7211972
IUPAC Name N-(4-methyl-1,3-thiazol-2-yl)-2-(6-phenylpyridazin-3-yl)sulfanylacetamide
SMILES CC1=CSC(=N1)NC(=O)CSC2=NN=C(C=C2)C3=CC=CC=C3

Specifications

Quantity 10 mg
Formula Weight 342.43
Percent Purity >99%
Chemical Name or Material VU 0240551
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.