Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VU 0365114, Tocris Bioscience™

Catalog No. 440450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-045-0 50 mg
44-041-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-045-0 Supplier R&D Systems Supplier No. 4404/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Positive allosteric modulator of M5 receptors

Selective positive allosteric modulator of M5 (EC50 values are 2.7 μM for human M5, and >30 μM for M1, M2, M3 and M4 receptors).

Chemical Identifiers

CAS 1208222-39-2
Molecular Formula C22H14F3NO3
Molecular Weight (g/mol) 397.353
InChI Key SPBGRXOPAXZSER-UHFFFAOYSA-N
Synonym 1-biphenyl-4-ylmethyl-5-trifluoromethoxy indoline-2,3-dione, 1-1,1'-biphenyl-4-ylmethyl-5-trifluoromethoxy-1h-indole-2,3-dione, 1-1,1'-biphenyl-4-ylmethyl-5-trifluoromethoxy indole-2,3-dione
PubChem CID 45281794
IUPAC Name 1-[(4-phenylphenyl)methyl]-5-(trifluoromethoxy)indole-2,3-dione
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CN3C4=C(C=C(C=C4)OC(F)(F)F)C(=O)C3=O

Specifications

Quantity 50 mg
Formula Weight 397.35
Percent Purity >98%
Chemical Name or Material VU 0365114
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.