Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VUF 11207 fumarate, Tocris Bioscience™

Catalog No. 478010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-801-0 10 mg
47-805-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-801-0 Supplier R&D Systems Supplier No. 4780/10
This item is not returnable. View return policy

Potent ACKR3 (CXCR7) agonist

Potent ACKR3 (CXCR7) chemokine receptor agonist (EC50 = 1.6 nM). Induces recruitment of β-arrestin to CXCR7 in HEK293-CXCR7 cells.

Chemical Identifiers

CAS 1378524-41-4
Molecular Formula C27H35FN2O4
Molecular Weight (g/mol) 470.59
InChI Key JVRRRKAXHMGUHZ-UHFFFAOYNA-N
Synonym e-n-3-2-fluorophenyl-2-methylallyl-3,4,5-trimethoxy-n-2-1-methylpyrrolidin-2-yl ethyl benzamide, vuf 11207 fumarate, qcr-79, jvrrrkaxhmguhz-xdjhfchbsa-n, n-2e-3-2-fluorophenyl-2-methylprop-2-en-1-yl-3,4,5-trimethoxy-n-2-1-methylpyrrolidin-2-yl ethyl benzamide
PubChem CID 59052285

Specifications

Quantity 10 mg
Formula Weight 586.65
Chemical Name or Material VUF 11207 fumarate
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.