Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VUF 8430 dihydrobromide, Tocris Bioscience™

Catalog No. 249410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-941-0 10 mg
24-945-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-941-0 Supplier R&D Systems Supplier No. 2494/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Potent, high affinity H4 agonist

High affinity (pKi = 7.5), potent histamine H4 receptor full agonist (pEC50 = 7.3). Displays moderate affinity for H3 receptors (pKi = 6.0) and weak partial agonist activity at H2 receptors.

Chemical Identifiers

CAS 100130-32-3
Molecular Formula C4H13Br2N5S
Molecular Weight (g/mol) 323.051
InChI Key GPWJSTKHQMIXCA-UHFFFAOYSA-N
Synonym vuf 8430 dihydrobromide, pseudourea, 2-2-guanidinoethyl-2-thio-, dihydrobromide, s-2-guanidylethyl-isothiourea, s,2-guanidinoethylisothiuronium bromide hydrobromide, carbamimidothioic acid, 2-aminoiminomethyl amino ethyl ester, dihydrobromide, 2-2-guanidinoethyl-2-thiopseudourea dihydrobromide, 2-aminoiminomethyl amino ethyl carbamimidothioic acid ester, carbamimidothioic acid,2-aminoiminomethyl amino ethyl ester, dihydrobromide 9ci, 2-carbamimidoylsulfanyl ethaneguanidine dihydrobromide, acmc-20m37r
PubChem CID 3063227
IUPAC Name 2-(diaminomethylideneamino)ethyl carbamimidothioate;dihydrobromide
SMILES C(CSC(=N)N)N=C(N)N.Br.Br

Specifications

Quantity 10 mg
Formula Weight 323.05
Percent Purity >98%
Chemical Name or Material VUF 8430 dihydrobromide
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.