Learn More
Streamlight™ Vulcan 180 Haz-Lo Class 1 Div 1 Intrinsically Safe Rechargeable Lantern
Catalog No. 19028334 Shop All Streamlight Products
Change view
Click to view available options
Color:
Orange
Yellow
Includes:
Quick Release Buckle Strap, 12 V DC Vehicle-mountable Direct Wire Charging Rack
Quick Release Buckle Strap, Charging Rack, AC/ DC Chargers
Quick Release Shoulder Strap, Rechargeable Battery
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
19-028-334 Orange Quick Release Shoulder Strap, Rechargeable Battery
19-028-331 Yellow Quick Release Shoulder Strap, Rechargeable Battery
19-028-332 Yellow Quick Release Buckle Strap, Charging Rack, AC/ DC Chargers
19-028-333 Yellow Quick Release Buckle Strap, 12 V DC Vehicle-mountable Direct Wire Charging Rack
19-028-335 Orange Quick Release Buckle Strap, Charging Rack, AC/ DC Chargers
19-028-336 Orange Quick Release Buckle Strap, 12 V DC Vehicle-mountable Direct Wire Charging Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-028-334 Supplier Streamlight™ Supplier No. 44330
Add to Cart
Add to Cart
Developed for use in hazardous locations on a portable scene light platform the Class 1, Div. 1 safety rated Vulcan 180 is lightweight and rechargeable. Combine the three extreme bright white LEDs with cordless operation, two ultra-bright blue taillights provide visibility in thick smoke.
  • Developed for use in hazardous locations on a portable scene light platform the Class 1, Div. 1 safety rated Vulcan 180 is lightweight and rechargeable
  • Combine the three extreme bright white LEDs with cordless operation to eliminate trip hazards while two ultra-bright blue taillights provide visibility in thick smoke
  • Features an articulating head that aims the beam where you need it; head locks in-place in forward facing position
  • Light Output: High for a bright, far-reaching beam: 400 lumens; 316m beam; runs 10 hours, Low for a less intense beam and longer run time: 130 lumens; 128m beam; runs 24 hours
  • Three LEDs produce a tight beam with optimum peripheral illumination
  • Two ultra-bright blue taillight LEDs make certain you can be seen even in thick smoke; can be programmed on/off or flashing
  • Optimized electronics provide regulated intensity
  • Large, multi-function, push-button switch easy to use with gloved hands
  • Rechargeable lithium Ion battery. Fully recharges in 15 hours with battery status indicator
Specifications
Light Source LED
Lighting Life 10 hr. (High), 24 hr. (Low)
Voltage 3.7 V
Battery Type Lithium-ion
Certifications/Compliance CLASS I, II, DIVISION 1, CLASS I, ZONE 0, GROUP IICGRPS A,B,C,D,E,F,G CLASS III; T-CODE: T4Meets applicable European Community Directives.Meets requirements of NFPA 1901-14.1.10.2 (2016) mounted in any position.CEC Compliant.
Color Orange
Includes Quick Release Shoulder Strap, Rechargeable Battery
Length (English) 7.43 in.
Material Nylon
Product Type Lantern/Scene Lighting
CLASS I, II, DIVISION 1, CLASS I, ZONE 0, GROUP IICGRPS A,B,C,D,E,F,G CLASS III; T-CODE: T4Meets applicable European Community Directives.Meets requirements of NFPA 1901-14.1.10.2 (2016) mounted in any position.CEC Compliant.