Learn More
Streamlight™ Vulcan 180 Haz-Lo Class 1 Div 1 Intrinsically Safe Rechargeable Lantern
Shop All Streamlight ProductsDescription
- Developed for use in hazardous locations on a portable scene light platform the Class 1, Div. 1 safety rated Vulcan 180 is lightweight and rechargeable
- Combine the three extreme bright white LEDs with cordless operation to eliminate trip hazards while two ultra-bright blue taillights provide visibility in thick smoke
- Features an articulating head that aims the beam where you need it; head locks in-place in forward facing position
- Light Output: High for a bright, far-reaching beam: 400 lumens; 316m beam; runs 10 hours, Low for a less intense beam and longer run time: 130 lumens; 128m beam; runs 24 hours
- Three LEDs produce a tight beam with optimum peripheral illumination
- Two ultra-bright blue taillight LEDs make certain you can be seen even in thick smoke; can be programmed on/off or flashing
- Optimized electronics provide regulated intensity
- Large, multi-function, push-button switch easy to use with gloved hands
- Rechargeable lithium Ion battery. Fully recharges in 15 hours with battery status indicator
Specifications
Specifications
| Light Source | LED |
| Lighting Life | 10 hr. (High), 24 hr. (Low) |
| Voltage | 3.7 V |
| Battery Type | Lithium-ion |
| Certifications/Compliance | CLASS I, II, DIVISION 1, CLASS I, ZONE 0, GROUP IICGRPS A,B,C,D,E,F,G CLASS III; T-CODE: T4Meets applicable European Community Directives.Meets requirements of NFPA 1901-14.1.10.2 (2016) mounted in any position.CEC Compliant. |
| Color | Orange |
| Includes | Quick Release Buckle Strap, Charging Rack, AC/ DC Chargers |
| Length (English) | 7.43 in. |
| Material | Nylon |
| Product Type | Lantern/Scene Lighting |
CLASS I, II, DIVISION 1, CLASS I, ZONE 0, GROUP IICGRPS A,B,C,D,E,F,G CLASS III; T-CODE: T4Meets applicable European Community Directives.Meets requirements of NFPA 1901-14.1.10.2 (2016) mounted in any position.CEC Compliant.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.