Learn More
Andwin Scientific VWR MINI SHAKER 120V, 50/60H
SDP
Supplier:  Andwin Scientific 12620938
Encompass_Preferred
WR Mini Shaker 120V 50/60Hz. Speed range: 100-1400Hz. Orbit: 3mm. Variable-speed microprocessor control. Independent LED display. Accommodates -10 to 60[degree]C 80% relative humidity non-condensing environment. Load: 3.6kg. Overall: 28Wx10.2Hx43.2Dcm.
Catalog No. NC1996650