Learn More
Selleck Chemical LLC VX-150 S1335-25mg
SDP
Supplier:  Selleck Chemical LLC S133525MG
Encompass_Preferred
VX-150 (EOS-62073) is an orally bioavailable pro-drug that rapidly converts into its active moiety which is a highly selective inhibitor of NaV1 8 relative to the other sodium channel subtypes ( 400-fold)
Catalog No. 50-217-3061
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.