Learn More
Medchemexpress LLC HY-13262 100mg , VX-809 Lumacaftor;VRT 826809 CAS:936727-05-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13262100MG
Encompass_Preferred
Medchemexpress, HY-13262 100mg VX-809 Lumacaftor;VRT 826809 CAS:936727-05-8 Formula:C24H18F2N2O5 EC50: 0.1 μM (CFTR) Purity:98% VX-809 is a CFTR corrector, which improves F508del-CFTR maturation with EC 50 of 0.1±0.1 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2682
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.