Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vybrant™ CFDA SE Cell Tracer Kit

Catalog No. V12883
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V12883 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V12883 Supplier Invitrogen™ Supplier No. V12883
  • $408.00
    $305.65 / Each of 1

    Save $102.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vybrant™ CFDA SE Cell Tracer Kit

Vybrant™ CFDA SE Cell Tracer Kit is used in fluorescent analyses as a long-term tracer of cells. This method is commonly used for in vitro and in vivo labeling of cells to determine whether or not a cell is proliferating.

  • Superior performance—allows for bright, uniform staining of cells
  • Long-term retention—dye is well-retained in cells for several days post stain
  • Non-toxic—minimal cytotoxicity for in vivo and in vitro assays
  • Simple, robust staining protocol

    Superior Fluorescent Staining
    Successful tracing of cells requires an extremely bright dye to distinguish fluorescently labeled cells from auto-fluorescence. The intense fluorescent staining provided by Vybrant™ CFDA SE Cell Tracer Kit enables the visualization of labeled cells for several days post stain, making it an ideal stain for cell tracing studies based on dye dilution.

    Long-Term Signal Retention
    Vybrant™ CFDA SE dye easily crosses the plasma membrane and covalently binds inside cells where the stable, well-retained fluorescent dye is designed to provide a consistent signal, even after several days in a cell culture environment.

    Non-Toxic
    Vybrant™ CFDA SE dye binds covalently to all free amines on the surface and inside of cells and shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells. Researchers have used CFDA SE labeling to show that transplantable hematopoietic cells proliferate in vitro in response to stimulation by a growth factor cocktail.

    Simple, Robust Staining Protocol
    The Vybrant™ CFDA SE Cell Tracer Kit contains convenient single-use vials of dry dye to permit small-scale experiments without preparing excess quantities of dye, thereby extending the shelf life of the dye not dissolved in solution. A stock solution is prepared by dissolving the contents of a vial in anhydrous DMSO prior to use. To stain 1 mL of cells in protein-free medium, 1μl of this stock solution is typically used. Cells should be stained for 20 minutes at room temperature with gentle agitation. A brief wash with complete medium will then quench any dye remaining in solution.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Contains 10 vials of 5(6)-CFDA, SE (CFSE, 500 μg per vial), and 1 vial of DMSO (1.3 mL). Store in freezer (-5 to -30°C).
    Excitation Wavelength Range 492⁄517
    Dye Type CFDA SE (CFSE)
    Format Tube(s), Slide(s)
    For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
    Quantity 1 Kit
    Detection Method Fluorescence
    Form Solid
    Product Line Vybrant
    Shipping Condition Room Temperature
    Solubility DMSO (Dimethylsulfoxide)
    Label Type Other Labels or Dyes
    Product Type Tracer
    Show More Show Less
    I'm trying to label my cells in suspension and track them over time by labeling with CFDA SE, but after labeling them they will no longer adhere to a surface (whereas unlabeled cells adhere well). I've tried to label at 10 and 20 µM. What is causing this?

    This dye will bind to proteins via primary amines. Too high of a concentration can lead to cell toxicity or unintended modification of cell function. The solution is to reduce the concentration and/or staining time, or to go with a non-protein-binding tracking reagent, such as Qtracker cell labeling reagents.

    I want to do a cell migration study for around 4 hours and need to fluorescently label the cells with a dye. What do you recommend?

    Calcein, AM and FDA (fluorescein diaceate) are examples of some dyes used for this application. Since these dyes are not incorporated or covalently attached to any cellular components, they may have a short retention time as some cell types may actively efflux the dye out of the cells. The CellTracker and CellTrace dyes include either a mild thiol-reactive chloromethyl group or amine-reactive succinnimidyl ester group to allow for covalent binding to cellular components, providing for better retention. As with any reagent, one should empirically determine retention times for the cell type used.

    I labeled my cells with Calcein, AM, but when I imaged the next day, there was no fluorescence from Calcein. Why?

    Calcein, AM is a good choice for cell tracking and as a general cytoplasmic stain. However, it doesn't bind to anything and may be actively pumped out of the cells within a couple hours, which is likely what happened. The retention of Calcein within live cells is dependent upon the inherent properties of the cell type and culture conditions.

    For long-term imaging, you may wish to consider a reactive cytoplasmic stains such as CFDA, SE or the CellTracker and CellTrace dyes.

    I want to track my cells with a nucleic acid stain, like DAPI or Hoechst dye. Do you recommend this?

    This is not recommended. When these stains bind to DNA and RNA, they may affect the normal function of the nucleic acids, disrupting transcription, as well as replication. Other reagents, such as CellTracker dyes or Qtracker reagents are more optimized for tracking without disrupting normal activity. If a nuclear label is still desired, though, and the cells are mammalian and non-hematopoietic, CellLight nuclear reagents can transiently transfect cells to express GFP or RFP on a nuclear-expressing protein for up to several days without affecting function.

    I want to track my cells over time, and you have a lot of options to choose from. How do I pick the right one?

    Please see this Web link (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/cell-tracking.html) to help you choose the right option for your application. Start by planning how long you want to track your cells, then consider the mechanism of binding. Calcein dyes are very uniform in label and are good for short-term cell migration, but may be rapidly effluxed from some cell types. Lipophilic cyanine dyes, such as DiI, DiO, and similar dyes label cell membranes, don’t disrupt function, and can last longer, but have the potential to cross to other cells if membranes fuse. They are also lost upon permeabilization. CellTracker dyes are better for longer-term labeling, as they possess a mildly reactive chloromethyl moiety that allows covalent binding to cellular components. CFDA SE also covalently binds to cellular components. With all the reagents, their retention within cells is dependent upon the rate of cell division and the inherent properties of the cell (active efflux, membrane and protein turnover rates, etc.) and reagents that allow for covalent attachment exhibit longer retention than those that do not.

    The longest-lasting and brightest options are the Qtracker reagents, which are taken up through endocytosis. These are so bright individual quantum dots can be detected, and are also robust enough to survive not only fixation and permeabilization, but even the heat and solvents used in paraffin processing.

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.