Learn More
Invitrogen™ Vybrant™ FLICA Caspase Apoptosis Assay Kits for flow cytometry
Description
Quickly detect caspase-mediated apoptosis by flow cytometry with Vybrant FAM caspase assay kits for poly-caspases and caspases 3 and 7. The Vybrant FAM caspase assay kits use fluorescent inhibitor of caspases (FLICA) methodology to detect and report caspase activity. Specifically, these caspase assay kits contain engineered FLICA substrates FAM-VAD-FMK (for poly-caspases), FAM-DEVD-FMK (for caspase-3/7), and FAM-LETD-FMK (for caspase-8). They also include Hoechst 33342 and propidium iodide stains, enabling simultaneous evaluation of membrane permeability and cell cycle by flow cytometry.
The Vybrant FAM caspase assay kits for flow cytometry employ a novel approach to detecting active caspases: the kits take advantage of FLICA methodology, which is an affinity label. This affinity label associates a fluoromethyl ketone (FMK) moiety, which can react covalently with a cysteine, with a caspase-specific amino acid sequence. Different amino acid recognition sequences are used to detect different caspases: valine-alanine-aspartic acid (VAD) for poly caspases (including caspase-1, -3, -4, -5, -6, -7, -8, and -9), aspartic acid-glutamic acid-valine-aspartic acid (DEVD) for caspase-3/7, and leucine-glutamic acid-threonine-aspartic acid (LETD) for caspase-8. A carboxyfluorescein (FAM) group is attached as a reporter.
The FLICA reagent is thought to interact with the enzymatic reactive center of an activated caspase via the recognition sequence, and then to attach covalently through the FMK moiety. The FLICA inhibitor is cell permeant and noncytotoxic. Unbound FLICA molecules diffuse out of the cell and are washed away; the remaining fluorescent signal is a direct measure of the amount of active caspase present at the time the inhibitor was added. The approximate excitation and emission peaks of the FLICA, propidium iodide, and Hoechst 33342 reagents are 488 nm/530 nm, 535 nm/6617 nm, and 350 nm/461 nm, respectively. The detection reagents included within the individual Vybrant FAM Poly Caspases Assay, Caspase-3 and -7 Assay, and Caspase-8 Assay apoptosis kits are 488/530 FAM-VAD-FMK reagent, 488/530 FAM-DEVD-FMK, and 488/530 FAM-LETD-FMK, respectively.
Specifications
Specifications
| Quantity | 25 Assays |
| Format | Tube |
| Product Type | Caspase Assay Kit |
| Content And Storage | 1 vial FAM-DEVD-FMK caspase-3 and -7 reagent (FLICA reagent, lyophilized solid), 1 vial of Hoechst 33342 (400 μL, 1 mM in water), 1 vial of PI (1 mL, 250 μg/mL in water), 1 vial of DMSO (0.5 mL), 1 bottle of apoptosis fixative solution (10% formaldehyde with methanol in PBS, 6 mL), and 1 bottle of 10x apoptosis wash buffer. Store in refrigerator (2–8°C) and protect from light. |
| Flow Cytometer Laser Lines | UV, 488 |
| Excitation/Emission | FAM-DEVD-FMD: 388/530 PI: 535/617 Hoechst 33342: 350/461 |
| Shipping Condition | Room Temperature |
| Product Line | Vybrant |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Flow Cytometer |
| Show More |
Frequently Asked Questions (FAQs)
FAM-DEVD-FMK FLICA reagent: 488/530 nm
Hoechst 33342: 350/461 nm
Propidium iodide: 535/617 nm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.