Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VZV Glycoprotein B Antibody [Alexa Fluor™ 405] - (Oka vaccine), Novus Biologicals™
SDP

Catalog No. NB560020
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB560020 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB560020 Supplier R&D Systems Supplier No. NBP343770AF405

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

VZV Glycoprotein B Polyclonal specifically detects VZV Glycoprotein B in Virus samples. It is validated for ELISA, Western Blot.

Specifications

Antigen VZV Glycoprotein B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA
Formulation 50 mM Sodium Borate
Gene Alias chicken pox, gB, gp140-62, gpII, ORF 31, varicella zoster, VZV
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Varicella Zoster Virus Glycoprotein B (Oka vaccine) (Uniprot#: Q4JR05)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.