Learn More
Sino Biological VZV VZV-gB Protein 100ug (Sino Biological)
Supplier:  Sino Biological 40958VNAS100
Encompass_Preferred
A DNA sequence encoding the Varicella-zoster virus (VZV) (strain Oka vaccine) Glycoprotein B (Q4JR05) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1180
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.