Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

W-5 hydrochloride, R&D Systems™
SDP

Catalog No. 036810 Shop All Novus Biologicals Products
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
03-681-0 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 03-681-0 Supplier R&D Systems Supplier No. 0368/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Calmodulin antagonist

Calmodulin antagonist (inhibits Ca2+-calmodulin dependent PDE with an IC50 of 240 μM).

Specifications

CAS 61714-25-8
Molecular Formula C16H23ClN2O2S
Quantity 10 mg
MDL Number MFCD00058043
Synonym w-5 hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hydrochloride, w-5, hydrochloride, n-6-aminohexyl naphthalene-1-sulfonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulphonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hcl, n-6-aminohexyl-1-naphthalenesulfonamide, hcl, acmc-209mvm, n-6-aminohexyl-1-naphthalenesulfonamide monohydrochlorde, 1-naphthalenesulfonamide, n-6-aminohexyl-, monohydrochlorde
InChI Key HOCSVIGHWPLMFC-UHFFFAOYSA-N
SMILES [H+].[Cl-].NCCCCCCNS(=O)(=O)C1=C2C=CC=CC2=CC=C1
IUPAC Name hydrogen N-(6-aminohexyl)naphthalene-1-sulfonamide chloride
Molecular Weight (g/mol) 342.88
PubChem CID 173829
Formula Weight 342.88
Percent Purity >98%
Product Line Tocris
Chemical Name or Material W-5 hydrochloride
Show More Show Less
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.