Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOMNI Scientific™ W.A.G Waste Anesthetic Gas Collection Filter

An activated charcoal filter developed to absorb 200 grams of halogenated agent.
Supplier: SOMNI Scientific™ WG15003
Description
- The W.A.G. filter can be utilized to passively and safely remove waste anesthetic gas at higher flowrates and from multiple sources
- Absorbs up to 200 grams of halogenated agent
- Use where evacuation to the outside is not possible
- Easily adapted for use with multiple sources of WAG
- Filter can be placed on a flat, stable surface or mounted using the W.A.G. Holder
- Inlet dimensions accomodate multiple tubing options (15 mm ID, 19/22 mm OD)
- Prevents unnecessary exposure when replaced regularly
Specifications
Not Autoclavable | |
White, Blue, Orange | |
Animal Inhalant Anesthesia | |
Single-use | |
4 filters | |
Round | |
Non-sterile | |
5 x 9 in. | |
5 in. | |
5 in. | |
9 in. | |
5 in. | |
12.7 cm | |
∼1 kg (2 lb.) |
200 g | |
Plastic | |
Veterinary | |
AMD-17106, AMD-17004, TT-17000, TT-17001, TT-17002, WG-15003, MW-17005, PH-4013 | |
Activated Carbon | |
One Size | |
5 x 5 x 9 in. | |
5 x 5 x 9 in. | |
12.7 cm | |
12.7 cm | |
18 cm | |
0.75 in. | |
19 mm, 22 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Veterinary Use Only
Spot an opportunity for improvement?Share a Content Correction