Learn More
Apexbio Technology LLC ω-Agatoxin IVA(Synonyms: Omega-Agatoxin IVA, ω-Aga IVA, Omega-Aga IVA, Agatoxin IVA, ω-Agatoxin-IVA, Aga IVA, ω-Aga-IVA), 10ug, CAS: 145017-83-0.
SDP
Supplier:  Apexbio Technology LLC B71920.01
Encompass_Preferred
ω-Agatoxin IVA (CAS 145017-83-0) is a peptide neurotoxin isolated originally from spider venom. It selectively inhibits voltage-gated P/Q-type calcium channels (Cav2.1), blocking calcium ion influx across neuronal membranes. By binding with high affinity to these calcium channels, ω-Agatoxin IVA suppresses neurotransmitter release at presynaptic terminals. Researchers routinely utilize ω-Agatoxin IVA to investigate neurotransmission, presynaptic calcium signaling, and the physiological or pathological roles of P/Q-type calcium channels in neuronal function.
Catalog No. 50-000-60081
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.