Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WAC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143930 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143930 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143930 Supplier Invitrogen™ Supplier No. PA5143930
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human A431 whole cell, human THP-1 whole cell, human Raji whole cell, human K562 whole cell, human U20S whole cell, human SK-OV-3 whole cell, rat brain tissue, rat PC-12 whole cell, mouse thymus tissue, mouse SP2/0 whole cell. ICC/IF: A431 cell. Flow: HepG2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The WW domain containing adaptor with coiled-coil protein (WAC) contains a WW domain that mediates protein-protein interactions and colocalizes with RNA splicing factor SC35. Further studies have indicated that WAC is a functional partner of the RNF20/40 complex that ubiquitinates Histone H2B, and that WAC regulates H2B ubiquitination. WAC targets RNF20/40 to associate with RNA polymerase II complex for H2B ubiquitination at active transcription sites. WAC-dependent transcription is also important for cell-cycle checkpoint activation in response to genotoxic stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WAC
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene Wac
Gene Accession No. Q924H7, Q9BTA9
Gene Alias 1110067P07Rik; A230035H12Rik; AI256735; AI256776; BM-016; DESSH; Kiaa1844; PRO1741; RGD1562407; Wac; WW domain containing adaptor with coiled-coil; WW domain-containing adapter protein with coiled-coil; WW domain-containing adaptor with coiled-coil; WW domain-containing protein 4; Wwp4
Gene Symbols Wac
Host Species Rabbit
Immunogen E.coli-derived human WAC recombinant protein (Position: Q459-V647).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225131, 307029, 51322
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.