Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Waffle Specimen Chucks for Leica, TBS, and Tanner Cryostats
SDP

Catalog No. 22445759 Shop All Mopec Products
Change view
Click to view available options
Color:
Blue
Green
White
Diameter (Metric):
30 mm
40 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Diameter (Metric)
22-445-759 Green 40 mm
22-445-766 Blue 30 mm
22-445-649 White 40 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-445-759 Supplier Mopec™ Supplier No. SP104WGN
Add to Cart
Add to Cart

Waffle Specimen Chucks are available in multiple sizes and colors.

  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats

Specifications

For Use With (Equipment) Leica, TBS, and Tanner Cryostats
Diameter (Metric) 40 mm
Color Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.