Learn More
Marketlab, Inc. Wall-Mount Metal Apparel/Coat Racks
Description
- Double hooks feature ball tips to help prevent damage to coats
- Easy to install; mounting hardware is included
- All Quickship furniture ships within 8 business days of order entry and credit approval
- Lead time may vary based on order volume; call for more details
Specifications
Specifications
| Width (English) | 36 in. |
| Size | One Size |
| Product Type | Coat Rack |
| Length (English) | 3.75 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.