Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Wall Mounted Arium pro Ultrapure Water System
GREENER_CHOICE

Catalog No. 14556016 Shop All Sartorius Products
Change view
Click to view available options
Conductivity:
100μS/cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conductivity
14-556-016 100μS/cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-556-016 Supplier Sartorius™ Supplier No. H2OBASICB
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Provides widest selection of features and options for customised configuration.

  • Modular units pre-assembled according to specific applications
  • Lowest TOC level, along with real-time monitoring function
  • Efficient removal of endotoxins, DNase and RNase
  • High Flexibility
  • Total ease of operation
  • Meets and exceeds the ASTM Type 1 standard, ensuring reproducibility of results
  • Applications: HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC analysis, PCR, Electrophoresis, Endotoxin analysis, Immunocytochemistry, Pure water for preparation of cell culture media, Pure water for production of monoclonal antibodies, Photometry
TRUSTED_SUSTAINABILITY

Specifications

Conductivity 100μS/cm
Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Distilled, DI, RO, EDI
Flow Rate 2 L/min.
Product Water Type Type 1
Remote Dispenser No
Resistivity 18.2 Mohm.cm
TOC Monitor with UV Intensity Monitoring < 5 ppb
Total Organic Carbon <5 ppb
DNase No
Mounting Wall
Particle Size 0.2μm
Silica <1000ppb
Daily Output 100 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type Arium pro Ultrapure Water System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.